accessibility menu, dialog, popup

UserName

CAS RN 14933-09-6

IUPAC Name: 3-[dimethyl(tetradecyl)azaniumyl]propane-1-sulfonate
Synonyms: myristyl sultaine
Molecular Weight (g/mol): 363.6
Molecular Formula: C19H41NO3S
InChi Key: BHATUINFZWUDIX-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (6)
  • (2)
Percent Purity 
  • (2)
  • (4)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results