missing translation for 'onlineSavingsMsg'
Learn More

Zirconium ICP Standard, 1000 ppm Zr in 3% HNO3, Ricca Chemical

Supplier:  Ricca Chemical Company PZR1KN500

 View more versions of this product

Catalog No. PZR1KN-500

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Zirconium ICP Standard
13826-66-9 , 7697-37-2 , 7732-18-5
94.0,4.22,0.26
1 mL = 1 mg Zr
H2N2O7Zr
< 2
Standard
[N+](=O)(O)[O-].[N+](=O)(O)[O-].O=[Zr]
233.247
83761
Natural Poly Bottle
1,000ppm Zr
97.0,4.39,0.27
Liquid
Colorless
Approximately 0°C
UJVRJBAUJYZFIX-UHFFFAOYSA-N
nitric acid;oxozirconium
500 mL

Chemical Identifiers

13826-66-9
233.247
83761
[N+](=O)(O)[O-].[N+](=O)(O)[O-].O=[Zr]
H2N2O7Zr
UJVRJBAUJYZFIX-UHFFFAOYSA-N
nitric acid;oxozirconium
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions