missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zirconium ICP Standard, 1000 ppm Zr in 3% HNO3, Ricca Chemical
$328.07 - $626.43
Chemical Identifiers
| CAS | 13826-66-9 |
|---|---|
| Molecular Formula | H2N2O7Zr |
| Molecular Weight (g/mol) | 233.247 |
| InChI Key | UJVRJBAUJYZFIX-UHFFFAOYSA-N |
| PubChem CID | 83761 |
| IUPAC Name | nitric acid;oxozirconium |
| SMILES | [N+](=O)(O)[O-].[N+](=O)(O)[O-].O=[Zr] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
PZR1KN-100
|
Ricca Chemical Company
PZR1KN100 |
100 mL | Natural LDPE Bottle |
N/A
|
|
||||
|
PZR1KN-500
|
Ricca Chemical Company
PZR1KN500 |
500 mL | Natural Poly Bottle |
N/A
|
|
||||
Chemical Identifiers
| 13826-66-9 | |
| 233.247 | |
| 83761 | |
| [N+](=O)(O)[O-].[N+](=O)(O)[O-].O=[Zr] |
| H2N2O7Zr | |
| UJVRJBAUJYZFIX-UHFFFAOYSA-N | |
| nitric acid;oxozirconium |
Specifications
| Natural LDPE Bottle | |
| 1,000ppm Zr | |
| 97.0,4.39,0.27 | |
| Liquid | |
| Colorless | |
| Approximately 0°C | |
| UJVRJBAUJYZFIX-UHFFFAOYSA-N | |
| nitric acid;oxozirconium | |
| 100 mL | |
| Zirconium ICP Standard |
| 13826-66-9 , 7697-37-2 , 7732-18-5 | |
| 94.0,4.22,0.26 | |
| 1 mL = 1 mg Zr | |
| H2N2O7Zr | |
| < 2 | |
| Standard | |
| [N+](=O)(O)[O-].[N+](=O)(O)[O-].O=[Zr] | |
| 233.247 | |
| 83761 |