missing translation for 'onlineSavingsMsg'
Learn More

Zirconium AA Standard, 1000 ppm Zr in 3% HNO3, Ricca Chemical

$115.40 - $312.67

Chemical Identifiers

CAS 13826-66-9
Molecular Formula H2N2O7Zr
Molecular Weight (g/mol) 233.247
InChI Key UJVRJBAUJYZFIX-UHFFFAOYSA-N
PubChem CID 83761
IUPAC Name nitric acid;oxozirconium
SMILES [N+](=O)(O)[O-].[N+](=O)(O)[O-].O=[Zr]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AZR1KN-100
SDS View Documents
Ricca Chemical Company
AZR1KN100
100 mL Natural LDPE Bottle N/A
Only null left
 
AZR1KN-500
SDS View Documents
Ricca Chemical Company
AZR1KN500
500 mL Natural Poly Bottle N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

13826-66-9
233.247
83761
[N+](=O)(O)[O-].[N+](=O)(O)[O-].O=[Zr]
H2N2O7Zr
UJVRJBAUJYZFIX-UHFFFAOYSA-N
nitric acid;oxozirconium

Specifications

Natural LDPE Bottle
93.0,4.34,0.26
97.0,4.52,0.27
Liquid
Colorless
Approximately 0°C
[N+](=O)(O)[O-].[N+](=O)(O)[O-].O=[Zr]
233.247
83761
13826-66-9 , 7697-37-2 , 7732-18-5
1,000ppm Zr
1 mL = 1 mg Zr
H2N2O7Zr
< 2
UJVRJBAUJYZFIX-UHFFFAOYSA-N
nitric acid;oxozirconium
100 mL
Zirconium AA Standard
Videos
SDS
Documents

Documents