missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zirconium AA Standard, 1000 ppm Zr in 3% HNO3, Ricca Chemical
Supplier: Ricca Chemical Company AZR1KN100
Specifications
| Zirconium AA Standard | |
| 13826-66-9 , 7697-37-2 , 7732-18-5 | |
| 1,000ppm Zr | |
| 1 mL = 1 mg Zr | |
| H2N2O7Zr | |
| < 2 | |
| UJVRJBAUJYZFIX-UHFFFAOYSA-N | |
| nitric acid;oxozirconium | |
| 100 mL |
| Natural LDPE Bottle | |
| 93.0,4.34,0.26 | |
| 97.0,4.52,0.27 | |
| Liquid | |
| Colorless | |
| Approximately 0°C | |
| [N+](=O)(O)[O-].[N+](=O)(O)[O-].O=[Zr] | |
| 233.247 | |
| 83761 |
Chemical Identifiers
| 13826-66-9 | |
| 233.247 | |
| 83761 | |
| [N+](=O)(O)[O-].[N+](=O)(O)[O-].O=[Zr] |
| H2N2O7Zr | |
| UJVRJBAUJYZFIX-UHFFFAOYSA-N | |
| nitric acid;oxozirconium |