missing translation for 'onlineSavingsMsg'
Learn More

Zinc carbonate basic, 97%, Zn >58.0%

Numéro de catalogue. aaa1459030
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAA1459030 250 g
AAA145900B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAA1459030 Fournisseur Thermo Scientific Chemicals Code fournisseur A1459030
Il en reste null

CAS: 5263-02-5 | C2H6O12Zn5 | 548.96 g/mol

Zinc carbonate basic is used as pharmaceutical intermediate and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc carbonate basic is used as pharmaceutical intermediate and in chemical research.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Identifiants chimiques

CAS 5263-02-5
Molecular Formula C2H6O12Zn5
Molecular Weight (g/mol) 548.96
MDL Number MFCD00011519
InChI Key UOURRHZRLGCVDA-UHFFFAOYSA-D
Synonym Zinc subcarbonate; Zinc carbonate hydroxide
IUPAC Name pentazinc(2+) hexahydroxide dicarbonate
SMILES [OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Zn++].[Zn++].[Zn++].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O

Spécifications

Quantity 250 g
Solubility Information Insoluble in water.
Formula Weight 548.97
Percent Purity 97%
Assay Zn >58.0%
Chemical Name or Material Zinc carbonate basic
Hazard Category H315-H319
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 226-076-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only