missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Zenix™ SEC-150 Gel Filtration HPLC Column, 3 μm Particle Size, Stainless Steel

Like their 5 micron siblings, 3 micron Zenix™ SEC columns feature a hydrophilic surface layer to prevent the sample from interacting with the underlying silica.

$3175.72 - $3602.82

Specifications

Length (Metric) 30 cm
Particle Size 3 μm
pH 2.0 to 8.5
Pore Size 150 Å
Stationary Phase Hydrophilic Polymer
View More Specs

Products 4
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
111005958
View Documents
MilliporeSigma™ Supelco™
Z777012
1 mm
Each for $3,602.82
Only null left
 
111005959
View Documents
MilliporeSigma™ Supelco™
Z777014
2.1 mm
Each for $3,602.82
Only null left
 
111006061
View Documents
MilliporeSigma™ Supelco™
Z777016
4.6 mm
Each for $3,175.72
Only null left
 
111005960
View Documents
MilliporeSigma™ Supelco™
Z777018
7.8 mm
Each for $3,602.82
Only null left
 
Description

Description

Select 100, 300 or 500 Å pores based on the size of your sample molecules, whether you are analyzing peptides, proteins, mAbs, ADCs, fusion proteins, etc. in quality control or research.
Specifications

Specifications

30 cm
2.0 to 8.5
Hydrophilic Polymer
Hydrophilic Polymer Phase/Spherical Silica
Stainless Steel
3 μm
150 Å
Zenix
Size Exclusion
SEC HPLC
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

Zenix is a registered trademark of Sepax Technologies