missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Zenix™ SEC-150 Gel Filtration HPLC Column, 3 μm Particle Size, Stainless Steel
Description
Select 100, 300 or 500 Å pores based on the size of your sample molecules, whether you are analyzing peptides, proteins, mAbs, ADCs, fusion proteins, etc. in quality control or research.
Specifications
Specifications
| Diameter (Metric) | 2.1 mm |
| Length (Metric) | 30 cm |
| Particle Size | 3 μm |
| pH | 2.0 to 8.5 |
| Pore Size | 150 Å |
| Stationary Phase | Hydrophilic Polymer |
| Product Line | Zenix |
| Matrix | Hydrophilic Polymer Phase/Spherical Silica |
| Phase | Size Exclusion |
| Material | Stainless Steel |
| Show More |
Zenix is a registered trademark of Sepax Technologies