Learn More
MilliporeSigma™ Supelco™ Zenix™ SEC-150 Gel Filtration HPLC Column, 3 μm Particle Size, Stainless Steel
Like their 5 micron siblings, 3 micron Zenix™ SEC columns feature a hydrophilic surface layer to prevent the sample from interacting with the underlying silica.
Supplier:  MilliporeSigma™ Supelco™ Z777012
 View more versions of this product
Catalog No. 111005958
Only null left
Explore available promotions
Description
Description
Select 100, 300 or 500 Å pores based on the size of your sample molecules, whether you are analyzing peptides, proteins, mAbs, ADCs, fusion proteins, etc. in quality control or research.
Specifications
Specifications
1 mm
3 μm
150 Å
Zenix
Size Exclusion
SEC HPLC
30 cm
2.0 to 8.5
Hydrophilic Polymer
Hydrophilic Polymer Phase/Spherical Silica
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Zenix is a registered trademark of Sepax Technologies