missing translation for 'onlineSavingsMsg'
Learn More

Yttrium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLY23X

 View more versions of this product

Catalog No. PLY2-3X


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Y10,000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Yttrium Standard
Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
MFCD00011473
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
RUDFQVOCFDJEEF-UHFFFAOYSA-N
5% HNO3
225.81
Assurance
Triple-leached LDPE bottle
1314-36-9 , 7732-18-5 , 7697-37-2
O3Y2
AA, ICP
Certified Reference Material
[O--].[O--].[O--].[Y+3].[Y+3]
diyttrium(3+) trioxidandiide
500 mL

Chemical Identifiers

1314-36-9
225.81
RUDFQVOCFDJEEF-UHFFFAOYSA-N
[O--].[O--].[O--].[Y+3].[Y+3]
O3Y2
MFCD00011473
diyttrium(3+) trioxidandiide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Causes skin irritation, serious eye irritation

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only