missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Yttrium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$428.78 - $1025.91
Chemical Identifiers
| CAS | 1314-36-9 |
|---|---|
| Molecular Formula | O3Y2 |
| Molecular Weight (g/mol) | 225.81 |
| MDL Number | MFCD00011473 |
| InChI Key | RUDFQVOCFDJEEF-UHFFFAOYSA-N |
| IUPAC Name | diyttrium(3+) trioxidandiide |
| SMILES | [O--].[O--].[O--].[Y+3].[Y+3] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Y | 10,000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
| 1314-36-9 | |
| 225.81 | |
| RUDFQVOCFDJEEF-UHFFFAOYSA-N | |
| [O--].[O--].[O--].[Y+3].[Y+3] |
| O3Y2 | |
| MFCD00011473 | |
| diyttrium(3+) trioxidandiide |
Specifications
| Triple-leached LDPE bottle | |
| 1314-36-9 , 7732-18-5 , 7697-37-2 | |
| O3Y2 | |
| AA, ICP | |
| Certified Reference Material | |
| [O--].[O--].[O--].[Y+3].[Y+3] | |
| diyttrium(3+) trioxidandiide | |
| 125 mL | |
| Yttrium Standard |
| Assurance Certificate of Analysis reports up to 30 elements at ppb levels. | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| MFCD00011473 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| RUDFQVOCFDJEEF-UHFFFAOYSA-N | |
| 5% HNO3 | |
| 225.81 | |
| Assurance |
Safety and Handling
Causes skin irritation, serious eye irritation
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only