missing translation for 'onlineSavingsMsg'
Learn More

Yeast Extract, Powder, MP Biomedicals™

Catalog No. ICN19402791
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19402791 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19402791 Supplier MP Biomedicals Inc Supplier No. 0219402791

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Yeast extract From Yeast, Molecular Biology Reagent, Yellow-tan Powder

  • It is used as a bacteriological media component for a variety of microorganisms and molecular genetics applications.
  • Yeast extract powder is a vacuum dried autolyzed yeast extract produced from 100% Baker's yeast.
  • It is a concentrated source of soluble protein components, carbohydrates, and B Complex vitamins natural to yeast.

Chemical Identifiers

CAS 8013-01-2

Specifications

Color Tan-Yellow
Form Solid
Purity or Quality Grade Molecular Biology Grade
Quantity 1 kg
Chemical Name or Material Yeast Extract
EINECSNumber 232-387-9