missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont Industrial™ Y7112 High Visibility Jersey Glove

Catalog No. 18064012
Change view
Click to view available options
Chlorinated:
No
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chlorinated
18064012 No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18064012 Supplier Wells Lamont Industrial™ Supplier No. Y7112L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hi-vis orange jersey glove with microdots on palm for grip. Knit wrist to keep debris out.

The high visibility jersey glove is made from quality materials and provides comfort and protection while on the job. Microdots around the palm and thumb provide a grip on the job. The knit wrist prevents irritants from entering the glove.

  • The Y7112 is an ideal glove in industries such as construction, railroad, maintenance, warehousing, and oil and gas.

Specifications

Chlorinated No
Construction Cut and Sewn
Applications Industrial
Color Hi-vis Orange
Hand Specific
Material Cotton
Product Type Multi-activity Gloves
Size Large
Gender Unisex