missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylitol, 98.5%, Spectrum™ Chemical
$490.32 - $2,913.14
Chemical Identifiers
| CAS | 87-99-0 |
|---|---|
| Molecular Formula | C5H12O5 |
| Molecular Weight (g/mol) | 152.15 |
| InChI Key | HEBKCHPVOIAQTA-UHFFFAOYNA-N |
| IUPAC Name | pentane-1,2,3,4,5-pentol |
| SMILES | OCC(O)C(O)C(O)CO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18614697
|
Spectrum Chemical Mfg Cor
XY107500GM |
500 g | Amber Glass Bottle |
N/A
|
|
||||
|
18614695
|
Spectrum Chemical Mfg Cor
XY10725KG |
2.5 kg | Amber Glass Bottle |
N/A
|
|
||||
|
18614694
|
Spectrum Chemical Mfg Cor
XY10712KG |
12 kg | Poly Pail |
N/A
|
|
||||
|
18614696
|
Spectrum Chemical Mfg Cor
XY10750KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.