missing translation for 'onlineSavingsMsg'
Learn More

Xylitol, 98.5%, Spectrum™ Chemical

Catalog No. 18614697
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614697 500 g Poly Pail
18614695 2.5 kg Poly Pail
18614694 12 kg Poly Pail
18614696 50 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18614697 Supplier Spectrum Chemical Mfg Cor Supplier No. XY107500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 87-99-0
Molecular Formula C5H12O5
Molecular Weight (g/mol) 152.15
InChI Key HEBKCHPVOIAQTA-UHFFFAOYNA-N
IUPAC Name pentane-1,2,3,4,5-pentol
SMILES OCC(O)C(O)C(O)CO

Specifications

CAS Max % 1
Boiling Point 216°C (420.8°F)
Packaging Poly Pail
Quantity 500 g
Percent Purity 0.985
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Xylitol

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.