missing translation for 'onlineSavingsMsg'
Learn More

Xylitol, 98.5%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor XY10750KGBL

 View more versions of this product

Catalog No. 18614696

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

87-99-0
Fiber Drum
50 kg
OCC(O)C(O)C(O)CO
152.15
Reagent
1
C5H12O5
HEBKCHPVOIAQTA-UHFFFAOYNA-N
pentane-1,2,3,4,5-pentol
0.985

Chemical Identifiers

87-99-0
152.15
pentane-1,2,3,4,5-pentol
C5H12O5
HEBKCHPVOIAQTA-UHFFFAOYNA-N
OCC(O)C(O)C(O)CO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.