missing translation for 'onlineSavingsMsg'
Learn More

Xylitol, 98.5%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor XY10712KG

 View more versions of this product

Catalog No. 18614694

Only null left

Description

Description

Specifications

Specifications

87-99-0
Poly Pail
12 kg
OCC(O)C(O)C(O)CO
152.15
Reagent
1
C5H12O5
HEBKCHPVOIAQTA-UHFFFAOYNA-N
pentane-1,2,3,4,5-pentol
0.985

Chemical Identifiers

87-99-0
152.15
pentane-1,2,3,4,5-pentol
C5H12O5
HEBKCHPVOIAQTA-UHFFFAOYNA-N
OCC(O)C(O)C(O)CO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.