missing translation for 'onlineSavingsMsg'
Learn More

Xylenol Orange Indicator, 0.1% (w/v), Ricca Chemical

Indicator for EDTA Titrations

$163.87 - $302.07

Chemical Identifiers

CAS 3618-43-7
Molecular Formula C31H28N2Na4O13S
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
9386-4
View Documents
Ricca Chemical Company
93864
120 mL Natural Poly Bottle
Each for $163.87
Only null left
 
9386-16
View Documents
Ricca Chemical Company
938616
500 mL Natural Poly Bottle
Each for $302.07
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3618-43-7
C31H28N2Na4O13S

Specifications

3618-43-7 , 7732-18-5
0.1% W/V
0.1% (w/v) Aqueous Solution
Orange to Dark Purple
Xylenol Orange Indicator
C31H28N2Na4O13S
Indicator
Natural Poly Bottle
120 mL
Videos
Safety and Handling

Safety and Handling

ShelfLife : 30 days

SDS
Documents

Documents