missing translation for 'onlineSavingsMsg'
Learn More

Xylenol Orange Indicator, 0.1% (w/v), Ricca Chemical

Indicator for EDTA Titrations

Supplier:  Ricca Chemical Company 938616

 View more versions of this product

Catalog No. 9386-16


Only null left

Description

Description

Specifications

Specifications

Xylenol Orange Indicator
C31H28N2Na4O13S
Indicator
Natural Poly Bottle
500 mL
3618-43-7 , 7732-18-5
0.1% W/V
0.1% (w/v) Aqueous Solution
Orange to Dark Purple

Chemical Identifiers

3618-43-7
C31H28N2Na4O13S
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 30 days

SDS
Documents

Documents

Promotions

Promotions