missing translation for 'onlineSavingsMsg'
Learn More

Corning™ X‐LAB™ System

Corning™ X‐LAB™ System is designed to separate and harvest a programmable volume of target cell populations from blood and blood products without use of density gradient media, in a closed, sterile device, ensuring consistent, reliable, efficient, high-recovery results.

$6,979.83 - $67,937.81

Specifications

For Use With (Equipment) X-LAB System
View More Specs

Products 3
missing translation for 'productCode' missing translation for 'brand' Product Type Quantity Price Quantity & Availability  
missing translation for 'productCode' missing translation for 'brand' Product Type Quantity Price Quantity & Availability  
10320117
View Documents Promotion Details
Corning™
6917
Control Module 1/Pk., 1/Cs. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10320118
View Documents Promotion Details
Corning™
6918
Docking Station 1/Pk., 1/Cs. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10320123
View Documents Promotion Details
Corning™
6924
Disposable Cartridges 1/Pk., 2/Cs.
Case of 2 Each for $6,979.83
Only null left
 
Description

Description

  • Customizable process programming, automated separation of samples into three chambers, and highly sensitive infrared sensors for cell separation ensure consistent, reliable, efficient, high-recovery results.
  • Processing data can be transferred to X-SERIES™ DataTrak software to further support GMP processing, monitoring and reporting of information.
  • Flexible solution for efficient, reliable, and reproducible density gradient media free recovery of target cell populations from blood and blood products.
  • Automated, functionally closed, sterile system
  • Density gradient media free sedimentation-based harvest
  • Consistent, high recovery (>90%) of viable cells
  • Customized protocols
  • Process 40 mL to 240 mL
  • Programmable harvest volume of 3 mL to 40 mL
  • Average processing time of 35 minutes depending on source material
  • Barcode tracking of supplies and process steps electronically
  • Precise selection of target cells
  • Single-use, disposable cartridge
  • US class I medical devices
Specifications

Specifications

X-LAB System
Videos
SDS
Documents

Documents