missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ Whizard™ Defender™ 7 Cut-Resistant Gloves

Numéro de catalogue. 19167805c
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Size
19167805C Medium
19167805B Small
19167805D Large
19167805E X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. 19167805C Fournisseur Wells Lamont™ Code fournisseur 135486
Il en reste null

Whizard™ Defender™ 7 Cut-Resistant Gloves with high performance fibers and stainless steel combination offer ANSI Cut Level A7 cut resistance. 7-gauge gloves are ideal for handling sharp surfaces, food preparation, and meat and poultry processing.

Patented combination of high-performance fibers and stainless steel creates superior cut resistant properties, eliminating the high cost of lacerations.

  • Standard cuff
  • Ambidextrous
  • Prewashed and preshrunk
  • 7 gauge: ANSI Level A7
  • Made in the USA

Spécifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance FDA CFR 21, ASTM F2992-15, ASTM D3884-09
Coating Material Uncoated
Color White, Yellow
Cut Resistance Rating ANSI A5
Hand Ambidextrous
Latex Free Yes
Material Fibers, Stainless Steel Fiber
Product Type Cut-resistant Gloves
Size Medium
For Use With (Application) Heavy duty work
Glove Cuff Color Yellow
Glove Cuff Style Plain
Hem Color Blue
Model 135486
Series Whizard™ Defender™ 7
Yarn Gauge 7
Afficher plus de résultats Afficher moins de résultats