Learn More
DWK Life Sciences Wheaton™ Square Media Bottles with Tamper Evident Caps
Square/Octagonal Media Bottles made of PETG that dispalys clarity, toughness, chemical inertness and acts as gas barrier
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|
02924970
|
DWK Life Sciences
WPBGC0030S |
30 mL |
N/A
|
|
|||||
|
02924973
|
DWK Life Sciences
WPBGC0250S |
250 mL |
N/A
|
|
|||||
|
02924975
|
DWK Life Sciences
WPBGC0500S |
500 mL |
N/A
|
|
|||||
|
02924976
|
DWK Life Sciences
WPBGC1000S |
1000 mL |
N/A
|
|
|||||
|
02924978
|
DWK Life Sciences
WPBGC2000S |
2000 mL |
N/A
|
|
|||||
Description
- For storage of media, laboratory reagents and biological buffers
- Made of Eastman Tritan Eastar Copolyester PETG that is resin used for single use labware and medical devices
- With tamper evident blue polypropylene caps
- Molded graduation markings for reference filling
- Gamma irradiated sterilized to SAL 10-6
- Cleanroom packaged for sterile production with multi-barrier elastomer packaging
- Resin meets ISO 10993 and/or USP Class VI biocompatibility requirements and is Food Contact Status compliant
Specifications
| PETG | |
| Sterile | |
| Square |