Learn More
DWK Life Sciences Wheaton™ Square Media Bottles with Tamper Evident Caps
Square/Octagonal Media Bottles made of PETG that dispalys clarity, toughness, chemical inertness and acts as gas barrier
Supplier: DWK Life Sciences WPBGC1000S
Description
- For storage of media, laboratory reagents and biological buffers
- Made of Eastman Tritan Eastar Copolyester PETG that is resin used for single use labware and medical devices
- With tamper evident blue polypropylene caps
- Molded graduation markings for reference filling
- Gamma irradiated sterilized to SAL 10-6
- Cleanroom packaged for sterile production with multi-barrier elastomer packaging
- Resin meets ISO 10993 and/or USP Class VI biocompatibility requirements and is Food Contact Status compliant
Chemical Identifiers
| PETG | |
| Sterile | |
| Square |
Specifications
| 1000 mL | |
| PETG | |
| Sterile | |
| Square |