Learn More
DWK Life Sciences Wheaton™ Square Media Bottles with Tamper Evident Caps
Square/Octagonal Media Bottles made of PETG that dispalys clarity, toughness, chemical inertness and acts as gas barrier
Supplier:  DWK Life Sciences WPBGC0500S
 View more versions of this product
Catalog No. 02924975
Only null left
Description
Description
  • For storage of media, laboratory reagents and biological buffers
  • Made of Eastman Tritan Eastar Copolyester PETG that is resin used for single use labware and medical devices
  • With tamper evident blue polypropylene caps
  • Molded graduation markings for reference filling
  • Gamma irradiated sterilized to SAL 10-6
  • Cleanroom packaged for sterile production with multi-barrier elastomer packaging
  • Resin meets ISO 10993 and/or USP Class VI biocompatibility requirements and is Food Contact Status compliant
Specifications
Chemical Identifiers
PETG
Sterile
Square
Specifications
500 mL
PETG
Sterile
Square
Product Suggestions
Product Suggestions