Learn More
DWK Life Sciences Wheaton™ Square Media Bottles with Tamper Evident Caps
Catalog No. 02924978
Change view
Click to view available options
Capacity (Metric):
30 mL
250 mL
500 mL
1000 mL
2000 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02924978 2000 mL
02924970 30 mL
02924973 250 mL
02924975 500 mL
02924976 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02924978 Supplier DWK Life Sciences Supplier No. WPBGC2000S
Square/Octagonal Media Bottles made of PETG that dispalys clarity, toughness, chemical inertness and acts as gas barrier
  • For storage of media, laboratory reagents and biological buffers
  • Made of Eastman Tritan Eastar Copolyester PETG that is resin used for single use labware and medical devices
  • With tamper evident blue polypropylene caps
  • Molded graduation markings for reference filling
  • Gamma irradiated sterilized to SAL 10-6
  • Cleanroom packaged for sterile production with multi-barrier elastomer packaging
  • Resin meets ISO 10993 and/or USP Class VI biocompatibility requirements and is Food Contact Status compliant
Specifications
Capacity (Metric) 2000 mL
Material PETG
Sterility Sterile
Shape Square