Learn More
DWK Life Sciences Wheaton™ ABC Clear Glass Vial Convenience Packs
40% larger opening improves sample accessibility while reducing autosampler needle damage
$99.75 - $336.67
Specifications
Diameter (English) Outer 0.47 in.
Color Clear
Diameter (Metric) Outer 12 mm
Material Borosilicate Glass
Volume (Metric) 1.8 mL
View More Specs
Products 19
Catalog Number Mfr. No. Marking Spot Closure Color Septum Pre-slit Price Quantity & Availability  
Catalog Number Mfr. No. Marking Spot Closure Color Septum Pre-slit Price Quantity & Availability  
02912910
View Documents Promotion Details
DWK Life Sciences
W2251540104
No Blue White PTFE, Red Silicone No
Case of 100 Each for $229.15
Only null left
03410737
View Documents Promotion Details
DWK Life Sciences
W2251500804
No Blue Red PTFE, White Silicone Yes
Case of 100 Each for $326.12
Only null left
03410221
View Documents Promotion Details
DWK Life Sciences
W2251500801
No Black Red PTFE, White Silicone Yes
Pack of 100 Each for $216.94
Only null left
02912907
View Documents Promotion Details
DWK Life Sciences
W22515401
No Natural White PTFE, Red Silicone No
Case of 100 Each for $253.57
Only null left
03410220
View Documents Promotion Details
DWK Life Sciences
W22515008
No Natural Red PTFE, White Silicone Yes
Pack of 100 Each for $176.10
Only null left
02912908
View Documents Promotion Details
DWK Life Sciences
W2251540101
No Black White PTFE, Red Silicone No
Case of 100 Each for $229.15
Only null left
02912909
View Documents Promotion Details
DWK Life Sciences
W2251540103
No Red White PTFE, Red Silicone No
Case of 100 Each for $229.15
Only null left
02912917
View Documents Promotion Details
DWK Life Sciences
W2251550105
Yes Yellow White PTFE, Red Silicone No
Case of 100 Each for $284.32
Only null left
03410738
View Documents Promotion Details
DWK Life Sciences
W2251510804
Yes Blue Red PTFE, White Silicone Yes
Case of 100 Each for $336.67
Only null left
03410209
View Documents Promotion Details
DWK Life Sciences
W2251500201
No Black Red PTFE, White Silicone No
Pack of 100 Each for $157.88
Only null left
02912912
View Documents Promotion Details
DWK Life Sciences
W2251540107
No Green White PTFE, Red Silicone No
Case of 100 Each for $264.89
Only null left
03410214
View Documents Promotion Details
DWK Life Sciences
W22515003
No Natural Red PTFE, White Silicone, Red PTFE No
Pack of 100 Each for $152.45
Only null left
02912913
View Documents Promotion Details
DWK Life Sciences
W22515501
Yes Natural White PTFE, Red Silicone No
Case of 100 Each for $272.21
Only null left
03410211
View Documents Promotion Details
DWK Life Sciences
W2251500204
No Blue Red PTFE, White Silicone No
Pack of 100 Each for $157.88
Only null left
03410208
View Documents Promotion Details
DWK Life Sciences
W22515002
No Natural Red PTFE, White Silicone No
Pack of 100 Each for $157.88
Only null left
02912916
View Documents Promotion Details
DWK Life Sciences
W2251550104
Yes Blue White PTFE, Red Silicone No
Case of 100 Each for $289.93
Only null left
02912914
View Documents Promotion Details
DWK Life Sciences
W2251550101
Yes Black White PTFE, Red Silicone No
Case of 100 Each for $289.93
Only null left
03410213
View Documents Promotion Details
DWK Life Sciences
W2251500207
No Green Red PTFE, White Silicone No
Pack of 100 Each for $157.88
Only null left
03410218
View Documents Promotion Details
DWK Life Sciences
W22515006
No Natural Red PTFE No
Pack of 100 Each for $99.75
Only null left
Description
Description
  • Manufactured from clear Type I borosilicate glass
  • 12 x 32 mm screw top with 9 mm ABC screw cap
  • Available with and without writing patch
Specifications
Specifications
0.47 in.
12 mm
1.8 mL
Polypropylene
9 mm
Clear
Borosilicate Glass
1.25 in.
Screw Cap
32 mm