missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Grade 603 Cellulose Extraction Thimbles
Range in size and length to ensure optimal fit.. Whatman™ Grade 603 Cellulose Extraction Thimbles are for use with extracting pesticides from environmental samples and fat analysis in the food industry
$161.28 - $467.00
Specifications
| Description | 603 Standard Cellulose Extraction Thimbles made from high-quality cellulose available in a range of sizes. |
|---|---|
| Material | Cellulose |
| For Use With (Application) | To analyze fats and extract organic compounds |
| Grade | 603 |
| Max. Temperature (English) | 266°F |
| Catalog Number | Mfr. No. | Diameter (Metric) Inside | Length (Metric) | Wall Thickness | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Diameter (Metric) Inside | Length (Metric) | Wall Thickness | Price | Quantity & Availability | |||
|
09924218
|
Cytiva
10350217 |
25 mm | 80 mm | 1.5 mm |
Pack of 25 Each for $195.22
|
|
|||
|
09924223
|
Cytiva
10350225 |
28 mm | 60 mm | 1.5 mm |
Pack of 25 Each for $161.28
|
|
|||
|
09924220
|
Cytiva
10350220 |
26 mm | 60 mm | 1.5 mm |
Pack of 25 Each for $186.87
|
|
|||
|
09924221
|
Cytiva
10350223 |
27 mm | 80 mm | 1.5 mm |
Pack of 25 Each for $178.43
|
|
|||
|
09924226
|
Cytiva
10350234 |
30 mm | 80 mm | 1.5 mm |
Pack of 25 Each for $178.43
|
|
|||
|
09924229
|
Cytiva
10350238 |
33 mm | 60 mm | 1.5 mm |
Pack of 25 Each for $178.43
|
|
|||
|
09924228
|
Cytiva
10350236 |
30 mm | 100 mm | 1.5 mm |
Pack of 25 Each for $209.00
|
|
|||
|
09924234
|
Cytiva
10350245 |
33 mm | 118 mm | 1.5 mm |
Pack of 25 Each for $285.27
|
|
|||
|
09924231
|
Cytiva
10350241 |
33 mm | 90 mm | 1.5 mm |
Pack of 25 Each for $217.00
|
|
|||
|
09924236
|
Cytiva
10350247 |
33 mm | 130 mm | 1.5 mm |
Pack of 25 Each for $261.00
|
|
|||
|
09924237
|
Cytiva
10350250 |
33 mm | 205 mm | 1.5 mm |
Pack of 25 Each for $317.00
|
|
|||
|
09924238
|
Cytiva
10350252 |
34 mm | 130 mm | 1.5 mm |
Pack of 25 Each for $266.00
|
|
|||
|
09924241
|
Cytiva
10350261 |
40 mm | 85 mm | 1.5 mm |
Pack of 25 Each for $267.00
|
|
|||
|
09924240
|
Cytiva
10350255 |
35 mm | 150 mm | 1.5 mm |
Pack of 25 Each for $316.00
|
|
|||
|
09924244
|
Cytiva
10350273 |
48 mm | 145 mm | 2 mm |
Pack of 25 Each for $435.28
|
|
|||
|
09927259
|
Cytiva
10350306 |
22 mm | 60 mm | 2 mm |
Pack of 25 Each for $247.00
|
|
|||
|
09927258
|
Cytiva
10350275 |
44 mm | 230 mm | 2 mm |
Pack of 25 Each for $467.00
|
|
|||
|
09924232
|
Cytiva
10350242 |
33 mm | 94 mm | 1.5 mm |
Pack of 25 Each for $237.11
|
|
|||
|
09924245
|
Cytiva
10350274 |
48 mm | 200 mm | 2 mm |
Pack of 25 Each for $418.00
|
|
|||
|
09924215
|
Cytiva
10350211 |
22 mm | 80 mm | 1.5 mm |
Pack of 25 Each for $202.13
|
|
|||
Description
Range in size and length to ensure optimal fit.. Whatman™ Grade 603 Cellulose Extraction Thimbles are for use with extracting pesticides from environmental samples and fat analysis in the food industry
Pure Cellulose Thimbles
- Made from pure cotton cellulose
- For extraction of pesticides from environmental samples, fat analysis in the food industry
- Single-thickness thimbles have wall thickness of approximately 1mm
- Double-thickness thimbles have ∽2mm wall thickness
- For high-temperature extractions up to 550°C (1022°F). Also for using solvents incompatible with cellulose thimbles
Specifications
| 603 Standard Cellulose Extraction Thimbles made from high-quality cellulose available in a range of sizes. | |
| To analyze fats and extract organic compounds | |
| 266°F | |
| Extraction Thimble |
| Cellulose | |
| 603 | |
| 130°C |