missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Grade 603 Cellulose Extraction Thimbles
Range in size and length to ensure optimal fit.. Whatman™ Grade 603 Cellulose Extraction Thimbles are for use with extracting pesticides from environmental samples and fat analysis in the food industry
Supplier: Cytiva 10350242
Description
Range in size and length to ensure optimal fit.. Whatman™ Grade 603 Cellulose Extraction Thimbles are for use with extracting pesticides from environmental samples and fat analysis in the food industry
Pure Cellulose Thimbles
- Made from pure cotton cellulose
- For extraction of pesticides from environmental samples, fat analysis in the food industry
- Single-thickness thimbles have wall thickness of approximately 1mm
- Double-thickness thimbles have ∽2mm wall thickness
- For high-temperature extractions up to 550°C (1022°F). Also for using solvents incompatible with cellulose thimbles
Chemical Identifiers
| 603 Standard Cellulose Extraction Thimbles made from high-quality cellulose available in a range of sizes. | |
| To analyze fats and extract organic compounds | |
| 266°F | |
| Extraction Thimble |
| Cellulose | |
| 603 | |
| 130°C |
Specifications
| 25 Pk. | |
| 33 mm | |
| 1.5 mm | |
| 603 | |
| 266°F | |
| Extraction Thimble |
| 603 Standard Cellulose Extraction Thimbles made from high-quality cellulose available in a range of sizes. | |
| Cellulose | |
| To analyze fats and extract organic compounds | |
| 94 mm | |
| 130°C |