missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ Reusable Full-Finger Knit Nylon Glove Liners

Catalog No. 189994591B
Change view
Click to view available options
Size:
Universal
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
189994591B Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 189994591B Supplier Wells Lamont™ Supplier No. M089

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reusable Full-Finger Knit Nylon Glove Liners are constructed of high-quality, 100% continuous filament nylon with a full finger design for those with severe glove sensitivities. Wear alone, or under nitrile gloves to help reduce perspiration.

  • Constructed of high-quality, 100% continuous filament nylon
  • Full-finger design provides protection for those with severe glove sensitivities
  • Form fitting for high-dexterity applications
  • Wear alone, or under nitrile gloves to help prevent perspiration
  • Universal size easily accommodates various hand sizes
  • Made in the USA

Specifications

Coating Material Uncoated
Color White
Disposable Reusable
Hand Ambidextrous
Laboratory/Cleanroom Use Yes
Latex Free Yes
Material Nylon
Product Type Glove Liners
Size Universal
Finger Style Full
For Use With (Application) Critical Environments, Cleanrooms, Electronics Assembly, and Inspection
Glove Cuff Style Knit
Series M08
Show More Show Less