missing translation for 'onlineSavingsMsg'
Learn More

Water with 0.05% Trifluoroacetic Acid (TFA), Optima LC/MS Grade, Fisher Chemical

Catalog No. LS1154
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LS1154 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LS1154 Supplier Thermo Fisher Scientific Supplier No. LS1154

missing translation for 'validMainframeMsg'

Ideal high purity mobile phase for LC-MS and UHPLC-UV applications, specifically protein and peptide separations.

  • Suitable for LC-MS and UHPLC-UV
  • Lower TFA content enhances separations by minimizing ion suppression
  • Pre-blended mobile phase simplifies preparation leaving more time to run and analyze samples
  • Tested for the absence of protease that could interfere with protein or peptide analysis
  • TFA is a common mobile phase additive and ion pair reagent in chromatography

Chemical Identifiers

CAS 7732-18-5

Specifications

Grade OPTIMA LC-MS
Quantity 4 L
Form Liquid
Reagent Type Water with 0.05% Trifluoroacetic Acid
Packaging Glass Bottle

Intended for research use only