Learn More
PolyCo™ VR™ Protective Gowns with Thumb Loops
Gowns provide greater coverage for applications where the threat of biological or chemical contamination exist.
Supplier: PolyCo™ 42400
Description
- Glove-to-sleeve protection
- Full-frontal coverage suitable for workplaces with biological or chemical hazards
- Slips on over workers' heads and ties securely in the back for a quick and easy fit
- Secures sleeves in place with thumb loop
- Durable 4 mil thickness
Specifications
| Controlled Environments Gown | |
| Non-sterile | |
| Full Sleeve with Thumb Loop |
| Ties | |
| Welded | |
| OSHA 29, CFR Part 1910.1030 |