missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Voltmeter

Ideal option for classroom electricity demonstrations.

Supplier:  United Scientific™ MVT003

 Afficher plus de versions de ce produit

Numéro de catalogue. S16046


Il en reste null

Description

Description

  • DC voltmeter mounted in a black plastic case.
  • Scale length of 75 mm, with ± 2.5% accuracy
  • Features 5-way binding post and an external zero-setting screw
  • Not protected against prolonged or significant overload
  • Stackable for convenient storage
Spécifications

Identifiants chimiques

Analog
Voltmeter
Classroom Demonstrations
1 Ea.

Spécifications

Classroom Demonstrations
Analog
3/10/15 V
Voltmeter
1 Ea.
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions