missing translation for 'onlineSavingsMsg'
Learn More
Learn More
United Scientific™ Voltmeter
Ideal option for classroom electricity demonstrations.
Supplier: United Scientific™ MVT005
Description
- DC voltmeter mounted in a black plastic case.
- Scale length of 75 mm, with ± 2.5% accuracy
- Features 5-way binding post and an external zero-setting screw
- Not protected against prolonged or significant overload
- Stackable for convenient storage
Identifiants chimiques
| Analog | |
| Voltmeter | |
| Classroom Demonstrations | |
| 1 Ea. |
Spécifications
| Classroom Demonstrations | |
| Analog | |
| 10 V |
| Voltmeter | |
| 1 Ea. |