missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Viral Production Medium, AGT™
Gibco Viral Production Medium, AGT, is the dry-format version of CTS Viral Production Medium, a chemically defined, serum-free, protein-free, animal origin-free, phenol red-free medium.
$328.68 - $12,350.40
Spécifications
| Cell Line | Viral Production Cells 2.0 |
|---|---|
| Cell Type | HEK 293 cells |
| Concentration | 25.6 g/L |
| Form | Powder (AGT) |
| Product Type | AAV Production Medium |
| Numéro de catalogue | Numéro du manufacturier. | Reconstituted Volume | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Reconstituted Volume | Prix | Quantity & Availability | |||||
|
A5147101
|
Gibco™
A5147101 |
1 L |
chaque for $328.68
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
A5147102
|
Gibco™
A5147102 |
10 L |
chaque for $2,514.90
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
A5147103
|
Gibco™
A5147103 |
50 L |
chaque for $9,877.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
A5147104
|
Gibco™
A5147104 |
100 L |
N/A
|
N/A | |||||
| Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
Description
- Gibco Viral Production Medium, AGT is the dry-format version of CTS Viral Production Medium, a chemically defined, serum-free, protein-free, animal origin-free, phenol red-free medium
- Medium is part of the CTS AAV-MAX Helper-Free AAV Production System for high-titer adeno-associated virus (AAV) production
- The CTS AAV-MAX Helper-Free AAV Production System is a scalable, high-titer platform for suspension AAV production with research-grade and GMP versions to enable seamless transition from research to commercial manufacturing
- Other components of the CTS AAV-MAX system include CTS Viral Production Cells 2.0, CTS AAV-MAX Transfection Kit, and CTS Viral-Plex Complexation Buffer.
Features of Viral Production Medium, AGT, include:
- Supports growth and transfection of CTS Viral Production Cells 2.0 from shake flask to bioreactor scales
- Helps enable production of high AAV titers, greater than 5 x 1010 vg/mL (unpurified)
- Simplifies transfer to manufacturing scale due to Advanced Granulation Technology (AGT) enabling auto-pH and simplified dissolution
- Formulated with GlutaMAX Supplement, no further supplementation required
- Manufactured in conformity with cGMP for medical devices, 21 CFR, Part 820, and certified to ISO 13485. Drug Master File and Regulatory Support File available upon request.
Advanced Granulation Technology
- Advanced Granulation Technology (AGT) helps simplify medium preparation at large scale with fast rehydration
- A just-add water protocol, and low dust generation, creates less pressure build-up during sterile filtration
- For more information on AGT media preparation at large scale, see Hydration and scale-up of AGT medium in HyPerforma and imPULSE Single-Use Mixers
Spécifications
| Viral Production Cells 2.0 | |
| 25.6 g/L | |
| AAV Production Medium | |
| Glucose, GlutaMAX | |
| AAV Production | |
| 6.9 to 7.5 | |
| Mammalian |
| HEK 293 cells | |
| Powder (AGT) | |
| Non-sterile | |
| No Phenol Red, No Antibiotics | |
| AAV-MAX | |
| Suspension Cell Culture |
Sécurité et manipulation
missing translation for 'shelfLife' : 12 Months