missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Viral Production Medium, AGT™

Catalog No. A5147102
Change view
Click to view available options
Reconstituted Volume:
1 L
10 L
50 L
100 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Reconstituted Volume
A5147102 10 L
A5147101 1 L
A5147103 50 L
A5147104 100 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A5147102 Supplier Gibco™ Supplier No. A5147102
Only null left

Gibco Viral Production Medium, AGT, is the dry-format version of CTS Viral Production Medium, a chemically defined, serum-free, protein-free, animal origin-free, phenol red-free medium.

  • Gibco Viral Production Medium, AGT is the dry-format version of CTS Viral Production Medium, a chemically defined, serum-free, protein-free, animal origin-free, phenol red-free medium
  • Medium is part of the CTS AAV-MAX Helper-Free AAV Production System for high-titer adeno-associated virus (AAV) production
  • The CTS AAV-MAX Helper-Free AAV Production System is a scalable, high-titer platform for suspension AAV production with research-grade and GMP versions to enable seamless transition from research to commercial manufacturing
  • Other components of the CTS AAV-MAX system include CTS Viral Production Cells 2.0, CTS AAV-MAX Transfection Kit, and CTS Viral-Plex Complexation Buffer.

Features of Viral Production Medium, AGT, include:

  • Supports growth and transfection of CTS Viral Production Cells 2.0 from shake flask to bioreactor scales
  • Helps enable production of high AAV titers, greater than 5 x 1010 vg/mL (unpurified)
  • Simplifies transfer to manufacturing scale due to Advanced Granulation Technology (AGT) enabling auto-pH and simplified dissolution
  • Formulated with GlutaMAX Supplement, no further supplementation required
  • Manufactured in conformity with cGMP for medical devices, 21 CFR, Part 820, and certified to ISO 13485. Drug Master File and Regulatory Support File available upon request.

Advanced Granulation Technology

  • Advanced Granulation Technology (AGT) helps simplify medium preparation at large scale with fast rehydration
  • A just-add water protocol, and low dust generation, creates less pressure build-up during sterile filtration
  • For more information on AGT media preparation at large scale, see Hydration and scale-up of AGT medium in HyPerforma and imPULSE Single-Use Mixers

Specifications

Cell Line Viral Production Cells 2.0
Cell Type HEK 293 cells
Concentration 25.6 g/L
Form Powder (AGT)
Product Type AAV Production Medium
Sterility Non-sterile
With Additives Glucose, GlutaMAX
Without Additives No Phenol Red, No Antibiotics
For Use With (Application) AAV Production
Product Line AAV-MAX
Quantity 10 L
pH 6.9 to 7.5
Reconstituted Volume 10 L
Content And Storage 10 L Securitainer, store at 2°C to 8°C protected from light
Culture Type Suspension Cell Culture
Expression System Mammalian
Show More Show Less