missing translation for 'onlineSavingsMsg'
Learn More

Vikan™ Stiff 19 in. Deck/Wall Scrub

Vikan™ Stiff 19 in. Deck/Wall Scrub is ideal for covering a lot of ground in food production facilities such as meat processing plants, bakeries, canneries, and dairy plants.

Supplier:  Vikan™ 70624

 Afficher plus de versions de ce produit

Numéro de catalogue. 17227810


Il en reste null

Description

Description

  • Wide scrubbing broom
  • Ideal for cleaning heavily soiled areas
  • Color-coded
Spécifications

Identifiants chimiques

Scrub Brush
Stiff
Polypropylene
FDA Compliant
19.1 in.
Large
Polyester
Yes
Non-sterile
Thread type Euro Hygienic
1 Ea.

Spécifications

Scrub Brush
Stiff
Polypropylene
FDA Compliant
Thread type Euro Hygienic
1 Ea.
Polyester
Yes
Non-sterile
Red
19.1 in.
Large
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions