missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ Furazolidone, MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | VETRANAL |
| CAS | 67-45-8 |
| Physical Form | Neat |
| Linear Formula | C8H7N3O5 |
| MDL Number | MFCD00010550 |
| Synonym | 3-(5-Nitrofurfurylideneamino)-2-oxazolidinone |
| Type | Analytical Standard |
| Quantity | 250 mg |
| Formula Weight | 225.16 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH