missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VETRANAL™ 2-Nitrobenzaldehyde semicarbazone-13C, 15N2, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Spécifications
Spécifications
| Product Line | VETRANAL |
| CAS | 957509-32-9 |
| Physical Form | Neat |
| Linear Formula | C713 CH8N215N2O3 |
| MDL Number | MFCD07370149 |
| Synonym | 2-NP-SCA-13 C, 15N2 |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 211.15 |
| Grade | Analytical Standard |
VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH