missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Vertical Electrophoresis Systems

Numéro de catalogue. p-143065
Change view
Click to view available options
Dimensions (L x W x H):
6.7 x 5.1 x 5.5 in.
Gel Dimensions:
10 x 10 cm
Volume (Metric):
300 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Dimensions (L x W x H) Gel Dimensions Volume (Metric)
FBVE101 6.7 x 5.1 x 5.5 in. 10 x 10 cm 300 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. FBVE101 Fournisseur Fisherbrand™ Code fournisseur P8 DS1 XMFIS

Choice of 10 x 10cm, 16 x 14cm, or 20 x 20cm gel size

Key Features

  • Systems are double-sided, allowing two protein gels to be run simultaneously
  • Gel assemblies don't need to be moved after the gel is set
  • Sliding clamp assemblies make system setup a snap and prevent leaks
  • Electrode configuration reduces run times
  • Increased electrode surface area provides exceptional field uniformity

Safety Features

  • Asymmetrical shape prevents mismatching of electrodes
  • Interlocking safety lid with attached power cords protect against electric shock

Spécifications

Gel Dimensions 10 x 10 cm
Includes Upper buffer chamber with cooling chamber, lower buffer chamber, safety lid (with attached power supply leads), gaskets, clamp assemblies, gel caster, four blank and two notched glass plates, two notched alumina plates, two 10-well, 0.8mm thick combs, four, 0.8mm thick side spacers
Volume (English) Buffer 10.14 oz.
Volume (Metric) 300 mL
Volume (Metric) Buffer 300 mL
Warranty 3 Years
Dimensions (L x W x H) 6.7 x 5.1 x 5.5 in.
Length (English) 6.7 in.
Length (English) Gel 3.93 in.
Length (Metric) 17 cm
Length (Metric) Gel 10 cm
Width (English) 5.1 in.
Width (English) Gel 3.93 in.
Width (Metric) 12.95 cm
Width (Metric) Gel 10 cm
Height (English) 5.5 in.
Height (Metric) 14 cm
Afficher plus de résultats Afficher moins de résultats