missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Zirconium (Zr) Standard for AAS 1000 ppm in 5% HCl/0.5% HF, Ricca Chemical
Description
- 1000 ppm in 5% HCl/0.5% HF
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| CAS Min % | 92.41%, 4.90%, 0.20% |
| CAS Max % | 96.19%, 5.10%, 0.20% |
| Specific Gravity | 1.03 |
| Physical Form | Liquid |
| UN Number | UN3264 |
| Melting Point | Approximately 0°C |
| Type | Single-element Standard Solution |
| Quantity | 500 mL |
| Grade | High Purity |
| Show More |