missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Zirconium (Zr) Standard for AAS 1000 ppm in 5% HCl/0.5% HF, Ricca Chemical

Catalog No. RV010089100
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010089100 100 mL
RV010089500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010089100 Supplier Ricca Chemical Company Supplier No. RV010089100N

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 1000 ppm in 5% HCl/0.5% HF
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 13520-92-8 , 7647-01-0 , 7732-18-5
Molecular Formula Cl2H16O9Zr
Molecular Weight (g/mol) 322.24
MDL Number MFCD00149898
InChI Key SDFOCNSOSARAKC-UHFFFAOYSA-L
PubChem CID 159678
IUPAC Name oxozirconium;octahydrate;dihydrochloride
SMILES O.Cl[Zr](Cl)=O

Specifications

Packaging Natural LDPE Bottle
CAS Min % 92.41%, 4.90%, 0.20%
CAS Max % 96.19%, 5.10%, 0.20%
Specific Gravity 1.03
Physical Form Liquid
UN Number UN3264
Melting Point Approximately 0°C
Quantity 100 mL
Grade High Purity