missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Zirconium (Zr) Standard for AAS 1000 ppm in 2% HNO3/0.5% HF, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Supplier:  Ricca Chemical Company RV010063500N

 View more versions of this product

Catalog No. RV010063500


Only null left

Description

Description

  • 1000 ppm in 2% HNO3/0.5% HF
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Specifications

Natural Poly Bottle
95.29%,1.96%,0.49%,0.26%
1.013
N2O7Zr
Approximately 0°C
[O-][N+](=O)O[Zr](=O)O[N+]([O-])=O
231.23
16211674
14985-18-3 , 7697-37-2 , 7664-39-3 , 7732-18-5
99.17%,2.04%,0.51%,0.28%
Liquid
MFCD00149896
JWBWFTANBHYHBC-UHFFFAOYSA-N
(nitrooxy)(oxo)zirconio nitrate
500 mL
High Purity

Chemical Identifiers

14985-18-3
231.23
JWBWFTANBHYHBC-UHFFFAOYSA-N
(nitrooxy)(oxo)zirconio nitrate
N2O7Zr
MFCD00149896
16211674
[O-][N+](=O)O[Zr](=O)O[N+]([O-])=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions