missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Zirconium (Zr) Standard for AAS 1000 ppm in 2% HNO3/0.5% HF, Ricca Chemical

Catalog No. RV010063100
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010063100 100 mL
RV010063500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010063100 Supplier Ricca Chemical Company Supplier No. RV010063100N

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 1000 ppm in 2% HNO3/0.5% HF
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 14985-18-3 , 7697-37-2 , 7664-39-3 , 7732-18-5
Molecular Formula N2O7Zr
Molecular Weight (g/mol) 231.23
MDL Number MFCD00149896
InChI Key JWBWFTANBHYHBC-UHFFFAOYSA-N
PubChem CID 16211674
IUPAC Name (nitrooxy)(oxo)zirconio nitrate
SMILES [O-][N+](=O)O[Zr](=O)O[N+]([O-])=O

Specifications

Packaging Natural LDPE Bottle
CAS Min % 95.29%, 1.96%, 0.49%, 0.26%
CAS Max % 99.17%, 2.04%, 0.51%, 0.28%
Specific Gravity 1.013
Physical Form Liquid
Melting Point Approximately 0°C
Quantity 100 mL
Grade High Purity