missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Tin (Sn) Standard for AAS 1000 ppm in 20% HCl, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Supplier:  Ricca Chemical Company RV010083500N

 View more versions of this product

Catalog No. RV010083500


Only null left
Explore available promotions

Description

Description

  • 1000 ppm in 20% HCl
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Specifications

Natural Poly Bottle
95.82%,1.96%,0.22%
1.112
Cl4Sn
Approximately 0°C
Cl[Sn](Cl)(Cl)Cl
260.51
24287
7646-78-8 , 7647-01-0 , 7646-78-8
99.74%,2.04%,0.22%
Liquid
UN1789
HPGGPRDJHPYFRM-UHFFFAOYSA-J
tetrachlorostannane
500 mL
High Purity

Chemical Identifiers

7646-78-8
260.51
24287
Cl[Sn](Cl)(Cl)Cl
Cl4Sn
HPGGPRDJHPYFRM-UHFFFAOYSA-J
tetrachlorostannane
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions