CAS RN 7646-78-8
Tin(IV) chloride, 1M solution in dichloromethane, AcroSeal™
CAS: 7646-78-8 | Cl4Sn | 260.52 g/mol
Tin(IV) chloride, 99.99% (metals basis)
CAS: 7646-78-8 Molecular Formula: Cl4Sn Molecular Weight (g/mol): 260.51 MDL Number: MFCD00011242 InChI Key: HPGGPRDJHPYFRM-UHFFFAOYSA-J Synonym: tin tetrachloride,tin iv chloride,stannane, tetrachloro,tin chloride,stannic chloride,tetrachlorotin,tin perchloride,tintetrachloride,tin iv tetrachloride,sncl4 PubChem CID: 24287 IUPAC Name: tetrachlorostannane SMILES: Cl[Sn](Cl)(Cl)Cl
Tin(IV) chloride, 99.999%, (trace metal basis)
CAS: 7646-78-8 Molecular Formula: Cl4Sn Molecular Weight (g/mol): 260.52 MDL Number: MFCD00011242 InChI Key: HPGGPRDJHPYFRM-UHFFFAOYSA-J Synonym: tin tetrachloride,tin iv chloride,stannane, tetrachloro,tin chloride,stannic chloride,tetrachlorotin,tin perchloride,tintetrachloride,tin iv tetrachloride,sncl4 PubChem CID: 24287 IUPAC Name: tetrachlorostannane SMILES: Cl[Sn](Cl)(Cl)Cl
Tin(IV) chloride, anhydrous, 98% (metals basis)
CAS: 7646-78-8 Molecular Formula: Cl4Sn Molecular Weight (g/mol): 260.51 MDL Number: MFCD00011242 InChI Key: HPGGPRDJHPYFRM-UHFFFAOYSA-J Synonym: tin tetrachloride,tin iv chloride,stannane, tetrachloro,tin chloride,stannic chloride,tetrachlorotin,tin perchloride,tintetrachloride,tin iv tetrachloride,sncl4 PubChem CID: 24287 IUPAC Name: tetrachlorostannane SMILES: Cl[Sn](Cl)(Cl)Cl
Tin(IV) chloride, 99%, anhydrous
CAS: 7646-78-8 Molecular Formula: Cl4Sn Molecular Weight (g/mol): 260.52 InChI Key: HPGGPRDJHPYFRM-UHFFFAOYSA-J Synonym: tin tetrachloride,tin iv chloride,stannane, tetrachloro,tin chloride,stannic chloride,tetrachlorotin,tin perchloride,tintetrachloride,tin iv tetrachloride,sncl4 PubChem CID: 24287 IUPAC Name: tetrachlorostannane SMILES: Cl[Sn](Cl)(Cl)Cl
Tin(IV) Chloride (ca. 1.0mol/L in Dichloromethane), TCI America™
CAS: 7646-78-8 Molecular Formula: Cl4Sn Molecular Weight (g/mol): 260.51 InChI Key: HPGGPRDJHPYFRM-UHFFFAOYSA-J Synonym: tin tetrachloride,tin iv chloride,stannane, tetrachloro,tin chloride,stannic chloride,tetrachlorotin,tin perchloride,tintetrachloride,tin iv tetrachloride,sncl4 PubChem CID: 24287 IUPAC Name: tetrachlorostannane SMILES: Cl[Sn](Cl)(Cl)Cl
VeriSpec™ Tin (Sn) Standard for AAS 1000 ppm in 20% HCl, Ricca Chemical
CAS: 7646-78-8 Molecular Formula: Cl4Sn Molecular Weight (g/mol): 260.51 InChI Key: HPGGPRDJHPYFRM-UHFFFAOYSA-J PubChem CID: 24287 IUPAC Name: tetrachlorostannane SMILES: Cl[Sn](Cl)(Cl)Cl