missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Tin (Sn) Standard for AAS 1000 ppm in 20% HCl, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

$249.47 - $439.50

Chemical Identifiers

CAS 7646-78-8
Molecular Formula Cl4Sn
Molecular Weight (g/mol) 260.51
InChI Key HPGGPRDJHPYFRM-UHFFFAOYSA-J
PubChem CID 24287
IUPAC Name tetrachlorostannane
SMILES Cl[Sn](Cl)(Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
RV010083100
View Documents
Ricca Chemical Company
RV010083100N
100 mL
Each for $249.47
Only null left
 
RV010083500
SDS View Documents
Ricca Chemical Company
RV010083500N
500 mL
Each for $439.50
Only null left
 
Description

Description

  • 1000 ppm in 20% HCl
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Chemical Identifiers

7646-78-8
260.51
24287
Cl[Sn](Cl)(Cl)Cl
Cl4Sn
HPGGPRDJHPYFRM-UHFFFAOYSA-J
tetrachlorostannane

Specifications

Natural LDPE Bottle
95.82%,1.96%,0.22%
1.112
Cl4Sn
Approximately 0°C
Cl[Sn](Cl)(Cl)Cl
260.51
24287
7646-78-8 , 7647-01-0 , 7646-78-8
99.74%,2.04%,0.22%
Liquid
UN1789
HPGGPRDJHPYFRM-UHFFFAOYSA-J
tetrachlorostannane
100 mL
High Purity
Videos
SDS
Documents

Documents