missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Tantalum Standard for ICP/MS 100 ppm in 2% HNO3/H, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Supplier:  Ricca Chemical Company RV010557100N

Catalog No. RV010557100


Only null left
Explore available promotions

Description

Description

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Specifications

Tantalium ICP-MS Standard
7783-71-3 , 7697-37-2 , 7732-18-5 , 7783-71-3
99.43%,2.04%,0.51%,0.02%
Liquid
Colorless
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Calibration Standard
YRGLXIVYESZPLQ-UHFFFAOYSA-I
pentafluorotantalum
100 mL
High Purity
Natural LDPE Bottle
95.53%,1.96%,0.49%,0.02%
1
F5Ta
Approximately 0°C
Miscible with water
Standard
F[Ta](F)(F)(F)F
275.94
82218

Chemical Identifiers

7783-71-3
275.94
82218
F[Ta](F)(F)(F)F
F5Ta
YRGLXIVYESZPLQ-UHFFFAOYSA-I
pentafluorotantalum
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions