accessibility menu, dialog, popup

UserName

CAS RN 7783-71-3

IUPAC Name: tantalum(5+) pentafluoride
Synonyms: tantalum pentafluoride
Molecular Weight (g/mol): 275.94
Molecular Formula: F5Ta
InChi Key: YRGLXIVYESZPLQ-UHFFFAOYSA-I
SMILES: [F-].[F-].[F-].[F-].[F-].[Ta+5]
Grade 
  • (8)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results