missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Tantalum Standard for ICP 10 g/L in 5% HNO3/HF, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Supplier:  Ricca Chemical Company RV010419100N

Catalog No. RV010419100


Only null left
Explore available promotions

Description

Description

  • 10 g/L in 5% HNO3/HF
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Specifications

Tantalum ICP Standard
7783-71-3 , 7697-37-2 , 7732-18-5 , 7783-71-3
93.31%,5.10%,2.04%,1.56%
Liquid
UN2922
<2
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Calibration Standard
YRGLXIVYESZPLQ-UHFFFAOYSA-I
pentafluorotantalum
100 mL
High Purity
Natural LDPE Bottle
89.65%,4.90%,1.96%,1.50%
1
F5Ta
Colorless
Approximately 0°C
Miscible with water
Standard
F[Ta](F)(F)(F)F
275.94
82218
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions