missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Tantalum Standard for ICP 1000 ppm in 5% HNO3/HF, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Supplier:  Ricca Chemical Company RV010318500N

 View more versions of this product

Catalog No. RV010318500


Only null left
Explore available promotions

Description

Description

  • 1000 ppm in 5% HNO3/HF
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Specifications

Natural Poly Bottle
91.97%,4.90%,0.98%,0.15%
1
F5Ta
Approximately 0°C
YRGLXIVYESZPLQ-UHFFFAOYSA-I
pentafluorotantalum
500 mL
High Purity
7783-71-3 , 7697-37-2 , 7732-18-5 , 7783-71-3
95.73%,5.10%,1.02%,0.15%
Liquid
UN2922
Standard
F[Ta](F)(F)(F)F
275.94
82218

Chemical Identifiers

7783-71-3
275.94
82218
F[Ta](F)(F)(F)F
F5Ta
YRGLXIVYESZPLQ-UHFFFAOYSA-I
pentafluorotantalum
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions